Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP5-6
Entrez Gene ID
440023
Gene Name
keratin associated protein 5-6
Gene Aliases
KRTAP5.6
Chromosome Location
Chr.11: 1697195 - 1697755 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012416.1 NP_001012416.1 1 - 1 497 96
GenBank mRNA AB126075.1 - 1 - 1 497 96
BC130399.1 - 1 - 1 469 96
BC133035.1 - 1 - 1 469 96

View Full Product Details