The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306159.1 | NP_001293088.1 | 9 - 10 | 923 | 130 |
NM_007082.4 | NP_009013.1 | 9 - 10 | 920 | 130 | |
XM_017003205.1 | XP_016858694.1 | 8 - 9 | 1673 | 130 | |
XM_017003215.1 | XP_016858704.1 | 8 - 9 | 1643 | 130 | |
GenBank mRNA | AF095350.1 | - | 9 - 10 | 848 | 130 |