Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SLX1B
- Entrez Gene ID
-
79008
- Gene Name
- SLX1 homolog B, structure-specific endonuclease subunit
- Gene Aliases
-
GIYD2
- Chromosome Location
-
Chr.16: 29454501 - 29458224 on Build GRCh38
- Gene Symbol
- SLX1A
- Entrez Gene ID
-
548593
- Gene Name
- SLX1 homolog A, structure-specific endonuclease subunit
- Gene Aliases
-
GIYD1
- Chromosome Location
-
Chr.16: 30193843 - 30197566 on Build GRCh38
View Full Product Details