The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001004470.1 | NP_001004470.1 | 2 - 3 | 200 | 135 |
GenBank mRNA | AB062101.1 | - | 2 - 3 | 200 | 135 |
AJ621583.1 | - | 2 - 3 | 200 | 135 | |
BC137102.1 | - | 2 - 3 | 332 | 135 | |
BC137103.1 | - | 2 - 3 | 332 | 135 | |
BC144472.1 | - | 2 - 3 | 332 | 135 |