Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCEAL1
Entrez Gene ID
9338
Gene Name
transcription elongation factor A like 1
Gene Aliases
SIIR, WEX9, p21, pp21
Chromosome Location
Chr.X: 103628720 - 103630953 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006639.1 NP_001006640.1 3 - 3 726 153
NM_001006640.1 NP_001006641.1 3 - 3 738 153
NM_004780.2 NP_004771.2 3 - 3 738 153
GenBank mRNA BC000809.1 - 3 - 3 691 153

View Full Product Details