Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A4
Entrez Gene ID
6583
Gene Name
solute carrier family 22 member 4
Gene Aliases
OCTN1
Chromosome Location
Chr.5: 132294384 - 132344206 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003059.2 NP_003050.2 10 - 10 1948 110
XM_006714675.3 XP_006714738.1 9 - 9 1355 110
XM_011543589.2 XP_011541891.1 8 - 8 1740 110
XM_017009776.1 XP_016865265.1 10 - 10 1996 110
GenBank mRNA AB007448.1 - 10 - 10 1929 110
BC028313.1 - 10 - 10 1948 110
Y09881.1 - 10 - 10 1941 110

View Full Product Details