Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CELF2-AS1
Entrez Gene ID
414196
Gene Name
CELF2 antisense RNA 1
Gene Aliases
C10orf31
Chromosome Location
Chr.10: 11316833 - 11344674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK097813.1 - 1 - 2 87 78

View Full Product Details