The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013653.2 | NP_001013675.1 | 1 - 2 | 772 | 114 |
GenBank mRNA | BC140911.1 | - | 1 - 2 | 741 | 114 |
CB160046.1 | - | 1 - 2 | 152 | 114 | |
DQ355157.1 | - | 1 - 2 | 708 | 114 | |
EU588721.1 | - | 1 - 2 | 163 | 114 |