The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001021.4 | NP_001012.1 | 2 - 3 | 275 | 88 |
NR_111943.1 | 1 - 2 | 481 | 88 | ||
NR_111944.1 | 2 - 3 | 275 | 88 | ||
GenBank mRNA | AK311951.1 | - | 2 - 3 | 183 | 88 |
BC009407.2 | - | 1 - 1 | 166 | 88 | |
BC019899.1 | - | 1 - 1 | 171 | 88 | |
BC020453.1 | - | 1 - 1 | 170 | 88 | |
BC022370.1 | - | 2 - 3 | 188 | 88 | |
BC049824.1 | - | 1 - 1 | 163 | 88 | |
BC062715.1 | - | 2 - 3 | 273 | 88 | |
BC070222.1 | - | 1 - 1 | 184 | 88 | |
BC071928.1 | - | 1 - 1 | 174 | 88 | |
BE905890.1 | - | 1 - 2 | 151 | 88 | |
BI597659.1 | - | 1 - 2 | 481 | 88 | |
BM457623.1 | - | 1 - 2 | 168 | 88 | |
DA310472.1 | - | 1 - 2 | 481 | 88 |