Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPIP5K1
Entrez Gene ID
9677
Gene Name
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location
Chr.15: 43533462 - 43590272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130858.2 NP_001124330.1 16 - 17 1851 73
NM_001130859.2 NP_001124331.1 15 - 16 1846 73
NM_001190214.1 NP_001177143.1 16 - 17 1851 73
NM_014659.5 NP_055474.3 15 - 16 1769 73
XM_005254803.1 XP_005254860.1 16 - 17 1851 73
XM_005254804.1 XP_005254861.1 16 - 17 1851 73
XM_006720786.2 XP_006720849.1 16 - 17 1851 73
XM_011522249.1 XP_011520551.1 16 - 17 1851 73
XM_011522250.1 XP_011520552.1 16 - 17 1954 73
XM_011522252.1 XP_011520554.1 15 - 16 1825 73
XM_011522254.1 XP_011520556.1 15 - 16 1721 73
XM_011522258.1 XP_011520560.1 10 - 11 1111 73
XM_011522259.1 XP_011520561.1 16 - 17 1851 73
XM_017022735.1 XP_016878224.1 15 - 16 2168 73
XM_017022736.1 XP_016878225.1 15 - 16 2136 73
XM_017022737.1 XP_016878226.1 15 - 16 1817 73
XM_017022738.1 XP_016878227.1 15 - 16 1765 73
XM_017022739.1 XP_016878228.1 15 - 16 1765 73
XM_017022740.1 XP_016878229.1 16 - 17 1954 73
XM_017022741.1 XP_016878230.1 15 - 16 2167 73
XM_017022742.1 XP_016878231.1 16 - 17 1959 73
XM_017022743.1 XP_016878232.1 15 - 16 1825 73
XM_017022744.1 XP_016878233.1 15 - 16 1766 73
XM_017022745.1 XP_016878234.1 16 - 17 1954 73
XM_017022746.1 XP_016878235.1 15 - 16 1825 73
XM_017022747.1 XP_016878236.1 15 - 16 1765 73
XM_017022748.1 XP_016878237.1 15 - 16 2167 73
XM_017022749.1 XP_016878238.1 16 - 17 1927 73
XM_017022750.1 XP_016878239.1 15 - 16 1768 73
XM_017022751.1 XP_016878240.1 15 - 16 1721 73
XM_017022752.1 XP_016878241.1 16 - 17 1851 73
XM_017022753.1 XP_016878242.1 15 - 16 1825 73
XM_017022754.1 XP_016878243.1 16 - 17 1959 73
XM_017022755.1 XP_016878244.1 15 - 16 1825 73
XM_017022756.1 XP_016878245.1 15 - 16 1763 73
XM_017022757.1 XP_016878246.1 15 - 16 1721 73
XM_017022758.1 XP_016878247.1 10 - 11 1111 73
XM_017022759.1 XP_016878248.1 15 - 16 1765 73
XM_017022760.1 XP_016878249.1 15 - 16 1825 73
GenBank mRNA AB002375.1 - 15 - 16 1773 73
AF502586.1 - 15 - 16 1741 73
AF502587.1 - 15 - 16 1773 73
AF502588.1 - 15 - 16 1741 73
AF502589.1 - 15 - 16 1773 73
AF543190.1 - 15 - 16 1723 73
AK307438.1 - 16 - 17 1815 73
BC050263.1 - 14 - 15 1797 73
BC057395.1 - 15 - 16 1797 73
BX647814.1 - 16 - 17 1806 73

View Full Product Details