Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPIP5K1
Entrez Gene ID
9677
Gene Name
diphosphoinositol pentakisphosphate kinase 1
Gene Aliases
HISPPD2A, IP6K, IPS1, VIP1, hsVIP1
Chromosome Location
Chr.15: 43533462 - 43590272 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130858.2 NP_001124330.1 4 - 5 545 104
NM_001130859.2 NP_001124331.1 3 - 4 540 104
NM_001190214.1 NP_001177143.1 4 - 5 545 104
NM_014659.5 NP_055474.3 3 - 4 463 104
XM_005254803.1 XP_005254860.1 4 - 5 545 104
XM_005254804.1 XP_005254861.1 4 - 5 545 104
XM_006720786.2 XP_006720849.1 4 - 5 545 104
XM_011522249.1 XP_011520551.1 4 - 5 545 104
XM_011522250.1 XP_011520552.1 4 - 5 648 104
XM_011522252.1 XP_011520554.1 3 - 4 519 104
XM_011522254.1 XP_011520556.1 3 - 4 415 104
XM_011522259.1 XP_011520561.1 4 - 5 545 104
XM_017022735.1 XP_016878224.1 3 - 4 862 104
XM_017022736.1 XP_016878225.1 3 - 4 830 104
XM_017022737.1 XP_016878226.1 3 - 4 511 104
XM_017022738.1 XP_016878227.1 3 - 4 459 104
XM_017022739.1 XP_016878228.1 3 - 4 459 104
XM_017022740.1 XP_016878229.1 4 - 5 648 104
XM_017022741.1 XP_016878230.1 3 - 4 861 104
XM_017022742.1 XP_016878231.1 4 - 5 653 104
XM_017022743.1 XP_016878232.1 3 - 4 519 104
XM_017022744.1 XP_016878233.1 3 - 4 460 104
XM_017022745.1 XP_016878234.1 4 - 5 648 104
XM_017022746.1 XP_016878235.1 3 - 4 519 104
XM_017022747.1 XP_016878236.1 3 - 4 459 104
XM_017022748.1 XP_016878237.1 3 - 4 861 104
XM_017022749.1 XP_016878238.1 4 - 5 621 104
XM_017022750.1 XP_016878239.1 3 - 4 462 104
XM_017022751.1 XP_016878240.1 3 - 4 415 104
XM_017022752.1 XP_016878241.1 4 - 5 545 104
XM_017022753.1 XP_016878242.1 3 - 4 519 104
XM_017022754.1 XP_016878243.1 4 - 5 653 104
XM_017022755.1 XP_016878244.1 3 - 4 519 104
XM_017022756.1 XP_016878245.1 3 - 4 457 104
XM_017022757.1 XP_016878246.1 3 - 4 415 104
XM_017022759.1 XP_016878248.1 3 - 4 459 104
XM_017022760.1 XP_016878249.1 3 - 4 478 104
GenBank mRNA AB002375.1 - 3 - 4 467 104
AF502586.1 - 3 - 4 435 104
AF502587.1 - 3 - 4 467 104
AF502588.1 - 3 - 4 435 104
AF502589.1 - 3 - 4 467 104
AF543190.1 - 3 - 4 417 104
AK307438.1 - 4 - 5 509 104
BC050263.1 - 2 - 3 491 104
BC057395.1 - 3 - 4 491 104
BX538022.1 - 3 - 4 491 104
BX647814.1 - 4 - 5 500 104

View Full Product Details