Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UQCRH
Entrez Gene ID
7388
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases
QCR6, UQCR8
Chromosome Location
Chr.1: 46303613 - 46316777 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006004.3 NP_005995.2 2 - 3 236 93
GenBank mRNA AK311967.1 - 2 - 3 132 93
BC001426.2 - 2 - 3 136 93
BC001934.1 - 2 - 3 141 93
BC015177.1 - 2 - 3 139 93
BC107703.1 - 2 - 3 142 93
BT007070.1 - 2 - 3 82 93
M36647.1 - 2 - 3 112 93
Y00764.1 - 2 - 3 118 93

Gene Symbol
UQCRHL
Entrez Gene ID
440567
Gene Name
ubiquinol-cytochrome c reductase hinge protein like
Gene Aliases
-
Chromosome Location
Chr.1: 15807162 - 15807699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089591.1 NP_001083060.1 1 - 1 132 93

View Full Product Details