Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCE2D
Entrez Gene ID
353141
Gene Name
late cornified envelope 2D
Gene Aliases
LEP12, SPRL1A
Chromosome Location
Chr.1: 152663411 - 152664659 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178430.3 NP_848517.1 2 - 2 426 170
GenBank mRNA AW300079.1 - 1 - 1 170

View Full Product Details