Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ATP7B
- Entrez Gene ID
-
540
- Gene Name
- ATPase copper transporting beta
- Gene Aliases
-
PWD, WC1, WD, WND
- Chromosome Location
-
Chr.13: 51932669 - 52012130 on Build GRCh38
View Full Product Details