The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024784.3 | NP_079060.1 | 2 - 2 | 1893 | 95 |
XM_011545256.2 | XP_011543558.1 | 2 - 2 | 1715 | 95 | |
GenBank mRNA | AK027045.1 | - | 2 - 2 | 1868 | 95 |
BC025249.1 | - | 2 - 2 | 1851 | 95 |