The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144997.5 | NP_659434.2 | 14 - 14 | 3063 | 81 |
XM_011523714.2 | XP_011522016.1 | 15 - 15 | 3114 | 81 | |
XM_011523716.1 | XP_011522018.1 | 13 - 13 | 3492 | 81 | |
XM_011523718.2 | XP_011522020.1 | 14 - 14 | 2998 | 81 | |
XM_011523721.2 | XP_011522023.1 | 14 - 14 | 2863 | 81 | |
XM_017024304.1 | XP_016879793.1 | 16 - 16 | 3283 | 81 | |
XM_017024305.1 | XP_016879794.1 | 16 - 16 | 3298 | 81 | |
XM_017024306.1 | XP_016879795.1 | 15 - 15 | 3287 | 81 | |
XM_017024307.1 | XP_016879796.1 | 15 - 15 | 3249 | 81 | |
XM_017024309.1 | XP_016879798.1 | 13 - 13 | 2838 | 81 | |
GenBank mRNA | AF517523.1 | - | 14 - 14 | 3014 | 81 |
BC015725.2 | - | 7 - 7 | 1763 | 81 |