Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHD
Entrez Gene ID
6007
Gene Name
Rh blood group D antigen
Gene Aliases
CD240D, DIIIc, RH, RH30, RHCED, RHDVA(TT), RHDel, RHPII, RHXIII, Rh4, RhDCw, RhII, RhK562-II, RhPI
Chromosome Location
Chr.1: 25272393 - 25330445 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127691.2 NP_001121163.1 7 - 7 1090 146
NM_001282867.1 NP_001269796.1 10 - 10 1374 146
NM_001282868.1 NP_001269797.1 8 - 8 1170 146
NM_001282869.1 NP_001269798.1 8 - 8 1224 146
NM_001282870.1 NP_001269799.1 9 - 9 1304 146
NM_001282871.1 NP_001269800.1 9 - 9 1394 146
NM_001282872.1 NP_001269801.1 10 - 10 1474 146
NM_016124.4 NP_057208.2 10 - 10 1378 146
GenBank mRNA AF187846.1 - 10 - 10 1316 146
AF510069.1 - 10 - 10 1359 146
AF510070.1 - 10 - 10 1359 146
AY751491.1 - 7 - 7 1028 146
AY751492.1 - 8 - 8 1108 146
AY751493.1 - 8 - 8 1162 146
AY751494.1 - 9 - 9 1242 146
AY751495.1 - 9 - 9 1332 146
AY751496.1 - 10 - 10 1412 146
BC139922.1 - 7 - 7 1078 146
DQ451878.1 - 10 - 10 1335 146
JQ424879.1 - 10 - 10 1374 146
X63094.1 - 10 - 10 1374 146
X63097.1 - 10 - 10 1332 146

View Full Product Details