Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
CYP4V2 | NM_207352.3 | 11 | 2450 | NP_997235.3 |
XM_005262935.3 | 11 | 2232 | XP_005262992.1 | |
XM_017008037.1 | 10 | 2208 | XP_016863526.1 | |
AK022114.1 | 5 | 1333 | ||
AK126473.1 | 12 | 2119 | BAC86562.1 | |
BC041839.1 | 4 | 1884 | AAH41839.1 | |
BX648537.1 | 5 | 1381 | ||
BX648730.1 | 6 | 6204 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3894090 | Chr4:186011626 - 186212421 on Build GRCh38 | Gain | CYP4V2 FLJ38576 TLR3 FAM149A |
nsv1028146 | Chr4:185582745 - 186417181 on Build GRCh38 | Gain | F11-AS1 F11 CYP4V2 SORBS2 KLKB1 FLJ38576 TLR3 FAM149A |
nsv4644 | Chr4:186185600 - 186219291 on Build GRCh38 | Insertion | CYP4V2 KLKB1 FLJ38576 |
nsv830172 | Chr4:186091611 - 186247270 on Build GRCh38 | Loss | CYP4V2 KLKB1 FLJ38576 FAM149A |
nsv508340 | Chr4:186138019 - 186215804 on Build GRCh38 | Deletion | CYP4V2 KLKB1 FLJ38576 FAM149A |
esv3603523 | Chr4:186176579 - 186214960 on Build GRCh38 | Gain | CYP4V2 FLJ38576 |
nsv461864 | Chr4:186085652 - 186335850 on Build GRCh38 | Gain | F11-AS1 F11 CYP4V2 KLKB1 FLJ38576 TLR3 FAM149A |
nsv596401 | Chr4:186151229 - 186445495 on Build GRCh38 | Loss | F11-AS1 F11 CYP4V2 KLKB1 FLJ38576 FAM149A |
esv3603517 | Chr4:186008703 - 186213065 on Build GRCh38 | Gain | CYP4V2 FLJ38576 TLR3 FAM149A |
dgv1052e214 | Chr4:186072002 - 186212619 on Build GRCh38 | Gain | CYP4V2 FLJ38576 TLR3 FAM149A |
nsv4642 | Chr4:186164028 - 186215260 on Build GRCh38 | Deletion | CYP4V2 FLJ38576 FAM149A |
esv3603525 | Chr4:186210472 - 186225457 on Build GRCh38 | Loss | CYP4V2 KLKB1 |