Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBC1D28
- Entrez Gene ID
-
254272
- Gene Name
- TBC1 domain family member 28
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 18634434 - 18644431 on Build GRCh38
- Gene Symbol
- TBC1D26
- Entrez Gene ID
-
353149
- Gene Name
- TBC1 domain family member 26
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 15732277 - 15744784 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK292544.1 |
- |
8 - 9 |
740 |
156 |
AK315847.1 |
- |
8 - 9 |
740 |
156 |
View Full Product Details