The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172732.1 | NP_001166203.1 | 3 - 3 | 1556 | 61 |
NM_001172739.1 | NP_001166210.1 | 3 - 3 | 1900 | 61 | |
NM_001172743.1 | NP_001166214.1 | 3 - 3 | 1903 | 61 | |
NM_021785.4 | NP_068557.3 | 2 - 2 | 1706 | 61 | |
NR_033348.1 | 3 - 3 | 1979 | 61 | ||
NR_033349.1 | 2 - 2 | 1785 | 61 | ||
XM_006724459.2 | XP_006724522.1 | 4 - 4 | 2055 | 61 | |
XM_006724460.1 | XP_006724523.1 | 3 - 3 | 1803 | 61 | |
XM_011545439.2 | XP_011543741.1 | 4 - 4 | 2084 | 61 | |
XM_011545440.2 | XP_011543742.1 | 4 - 4 | 2087 | 61 | |
XM_011545441.2 | XP_011543743.1 | 3 - 3 | 1890 | 61 | |
GenBank mRNA | AK056214.1 | - | 3 - 3 | 1799 | 61 |
AK298873.1 | - | 3 - 3 | 1517 | 61 | |
AK314631.1 | - | 2 - 2 | 1669 | 61 | |
BC027937.1 | - | 3 - 3 | 1859 | 61 | |
CK902458.1 | - | 1 - 1 | 61 |