Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO6
Entrez Gene ID
196527
Gene Name
anoctamin 6
Gene Aliases
BDPLT7, SCTS, TMEM16F
Chromosome Location
Chr.12: 45215987 - 45440404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025356.2 NP_001020527.2 15 - 16 2214 84
NM_001142678.1 NP_001136150.1 15 - 16 2061 84
NM_001142679.1 NP_001136151.1 15 - 16 2214 84
NM_001204803.1 NP_001191732.1 16 - 17 2277 84
XM_005268706.4 XP_005268763.1 15 - 16 2168 84
XM_005268707.3 XP_005268764.1 14 - 15 2088 84
GenBank mRNA AK126409.1 - 15 - 16 2295 84
AK302819.1 - 16 - 17 2071 84
AK303996.1 - 15 - 16 2145 84
AK310641.1 - 15 - 16 2181 84
AL832340.1 - 15 - 16 2061 84
BC098410.1 - 15 - 16 2081 84
BC136445.1 - 16 - 17 1970 84

View Full Product Details