The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144999.3 | NP_659436.1 | 3 - 4 | 698 | 64 |
GenBank mRNA | BC014109.2 | - | 3 - 4 | 596 | 64 |