Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EIF2S3
- Entrez Gene ID
-
1968
- Gene Name
- eukaryotic translation initiation factor 2 subunit gamma
- Gene Aliases
-
EIF2, EIF2G, EIF2gamma, eIF-2gA
- Chromosome Location
-
Chr.X: 24054948 - 24078810 on Build GRCh38
- Gene Symbol
- LOC255308
- Entrez Gene ID
-
255308
- Gene Name
- eukaryotic translation initiation factor 2 subunit gamma pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.12: 10505883 - 10508527 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC087847.1 |
- |
1 - 1 |
764 |
73 |
View Full Product Details