Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDRT15
Entrez Gene ID
146822
Gene Name
CMT1A duplicated region transcript 15
Gene Aliases
-
Chromosome Location
Chr.17: 14235856 - 14236833 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007530.1 NP_001007531.1 1 - 2 259 94

View Full Product Details