The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033655.3 | NP_387504.2 | 10 - 11 | 1889 | 98 |
GenBank mRNA | AB051501.2 | - | 10 - 11 | 1796 | 98 |
AF333769.2 | - | 10 - 11 | 1757 | 98 | |
AK056833.1 | - | 10 - 11 | 1889 | 98 | |
BC132737.1 | - | 10 - 11 | 1794 | 98 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201380.2 | NP_001188309.2 | 10 - 11 | 2060 | 98 |