Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRAMEF5
Entrez Gene ID
343068
Gene Name
PRAME family member 5
Gene Aliases
PRAMEF23, PRAMEF5L
Chromosome Location
Chr.1: 13254198 - 13263435 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013407.2 NP_001013425.2 4 - 4 1766 87

View Full Product Details