Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COLCA1
Entrez Gene ID
399948
Gene Name
colorectal cancer associated 1
Gene Aliases
C11orf92, CASC12, LOH11CR1F
Chromosome Location
Chr.11: 111293389 - 111305048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302644.1 NP_001289573.1 2 - 2 2652 144
NM_001302645.1 NP_001289574.1 2 - 2 2519 144
NM_001302646.1 NP_001289575.1 2 - 2 2690 144
NM_001302647.1 NP_001289576.1 2 - 2 2560 144
NM_001302648.1 NP_001289577.1 2 - 2 2525 144
NM_207429.3 NP_997312.1 2 - 2 2734 144
GenBank mRNA AB096245.1 - 2 - 2 2651 144
AK127703.1 - 1 - 1 377 144
CR936778.1 - 2 - 2 2649 144
KF728566.1 - 2 - 2 2652 144
KF728567.1 - 2 - 2 2519 144
KF728568.1 - 2 - 2 2734 144
KF728569.1 - 2 - 2 2652 144
KF728570.1 - 2 - 2 2690 144

View Full Product Details