The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321065.1 | NP_001307994.1 | 2 - 3 | 324 | 98 |
NM_001321066.1 | NP_001307995.1 | 3 - 4 | 386 | 98 | |
NM_001645.4 | NP_001636.1 | 2 - 3 | 260 | 98 | |
GenBank mRNA | AI310334.1 | - | 2 - 3 | 98 | |
AI885116.1 | - | 2 - 3 | 98 | ||
AK312036.1 | - | 2 - 3 | 141 | 98 | |
BC009698.2 | - | 1 - 2 | 83 | 98 | |
BC055093.1 | - | 3 - 4 | 312 | 98 | |
BT007142.1 | - | 1 - 2 | 61 | 98 | |
CR456907.1 | - | 1 - 2 | 61 | 98 | |
DA482306.1 | - | 3 - 4 | 386 | 98 | |
X00570.1 | - | 1 - 2 | 117 | 98 |