The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001480.2 | NP_001001480.2 | 1 - 1 | 1058 | 83 |
GenBank mRNA | AJ628247.1 | - | 3 - 3 | 1010 | 83 |