Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VWC2L
Entrez Gene ID
402117
Gene Name
von Willebrand factor C domain containing protein 2-like
Gene Aliases
-
Chromosome Location
Chr.2: 214411737 - 214575929 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080500.2 NP_001073969.1 2 - 3 520 108
GenBank mRNA AB374231.1 - 1 - 2 390 108
BC146903.1 - 2 - 3 520 108
BC146911.1 - 2 - 3 520 108
BC146931.1 - 2 - 3 520 108
BC146935.1 - 2 - 3 520 108
EU541473.1 - 1 - 2 393 108

View Full Product Details