Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR2B11
Entrez Gene ID
127623
Gene Name
olfactory receptor family 2 subfamily B member 11
Gene Aliases
-
Chromosome Location
Chr.1: 247451029 - 247451982 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004492.1 NP_001004492.1 1 - 1 810 83
GenBank mRNA BC137181.1 - 1 - 1 834 83
BC137205.1 - 1 - 1 834 83

View Full Product Details