Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CYP2D6
- Entrez Gene ID
-
1565
- Gene Name
- cytochrome P450 family 2 subfamily D member 6
- Gene Aliases
-
CPD6, CYP2D, CYP2D7AP, CYP2D7BP, CYP2D7P2, CYP2D8P2, CYP2DL1, CYPIID6, P450-DB1, P450C2D, P450DB1
- Chromosome Location
-
Chr.22: 42125531 - 42130881 on Build GRCh38
- Gene Symbol
- CYP2D7
- Entrez Gene ID
-
1564
- Gene Name
- cytochrome P450 family 2 subfamily D member 7 (gene/pseudogene)
- Gene Aliases
-
CYP2D, CYP2D7AP, CYP2D7P, CYP2D7P1, CYP2D@, P450C2D, P450DB1, RNA40057
- Chromosome Location
-
Chr.22: 42140203 - 42144575 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002570.3 |
|
7 - 8 |
1286 |
94 |
GenBank mRNA |
AY220845.1 |
- |
8 - 9 |
1348 |
94 |
X07619.1 |
- |
4 - 5 |
886 |
94 |
X07620.1 |
- |
3 - 4 |
266 |
94 |
X16866.1 |
- |
1 - 2 |
278 |
94 |
- Gene Symbol
- LOC101929829
- Entrez Gene ID
-
101929829
- Gene Name
- cytochrome P450 family 2 subfamily D member 6 pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_111920.1 |
|
7 - 8 |
1263 |
94 |
GenBank mRNA |
AB209492.1 |
- |
6 - 7 |
3442 |
94 |
M24499.1 |
- |
5 - 6 |
802 |
94 |
X16865.1 |
- |
5 - 6 |
808 |
94 |
View Full Product Details