The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001022.3 | NP_001013.1 | 4 - 5 | 726 | 153 |
NM_001321483.1 | NP_001308412.1 | 4 - 5 | 403 | 153 | |
NM_001321484.1 | NP_001308413.1 | 4 - 5 | 533 | 153 | |
NM_001321485.1 | NP_001308414.1 | 4 - 5 | 739 | 153 | |
XM_017027113.1 | XP_016882602.1 | 4 - 5 | 721 | 153 | |
GenBank mRNA | AK311786.1 | - | 4 - 5 | 389 | 153 |
BC000023.2 | - | 4 - 5 | 423 | 153 | |
BC007615.2 | - | 3 - 4 | 354 | 153 | |
BC017386.1 | - | 4 - 5 | 392 | 153 | |
BC018616.1 | - | 4 - 5 | 726 | 153 | |
BG398397.1 | - | 4 - 5 | 533 | 153 | |
BU603942.1 | - | 4 - 5 | 405 | 153 | |
CB124389.1 | - | 4 - 5 | 432 | 153 | |
CB125125.1 | - | 4 - 5 | 403 | 153 | |
M81757.1 | - | 4 - 5 | 376 | 153 |