The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001540.3 | NP_001531.1 | 2 - 3 | 581 | 145 |
GenBank mRNA | AB020027.1 | - | 2 - 3 | 451 | 145 |
AK296890.1 | - | 3 - 4 | 361 | 145 | |
AK311446.1 | - | 2 - 3 | 213 | 145 | |
AK311894.1 | - | 2 - 3 | 464 | 145 | |
AL050380.1 | - | 3 - 4 | 312 | 145 | |
BC000510.2 | - | 2 - 3 | 525 | 145 | |
BC012292.1 | - | 2 - 3 | 499 | 145 | |
BC012768.2 | - | 2 - 3 | 448 | 145 | |
BC014920.1 | - | 2 - 3 | 385 | 145 | |
BC073768.1 | - | 2 - 3 | 446 | 145 | |
BT019888.1 | - | 2 - 3 | 426 | 145 | |
CR407614.1 | - | 2 - 3 | 426 | 145 | |
CR536489.1 | - | 2 - 3 | 426 | 145 | |
FJ224323.1 | - | 2 - 3 | 533 | 145 | |
S74571.1 | - | 1 - 2 | 63 | 145 | |
U90906.1 | - | 2 - 3 | 533 | 145 | |
X16477.1 | - | 2 - 3 | 102 | 145 | |
X54079.1 | - | 2 - 3 | 478 | 145 |