Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG6
Entrez Gene ID
59285
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Gene Aliases
-
Chromosome Location
Chr.19: 53991149 - 54012666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145815.1 NP_665814.1 2 - 3 995 62
NR_102308.1 2 - 3 123 62
GenBank mRNA BX281606.1 - 2 - 3 123 62

View Full Product Details