The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022789.3 | NP_073626.1 | 1 - 2 | 534 | 76 |
NM_172314.1 | NP_758525.1 | 2 - 3 | 386 | 76 | |
GenBank mRNA | AF305200.1 | - | 1 - 2 | 534 | 76 |
AF458059.1 | - | 1 - 2 | 228 | 76 | |
AF461739.1 | - | 2 - 3 | 386 | 76 | |
AY359127.1 | - | 1 - 2 | 534 | 76 | |
BC069565.1 | - | 1 - 2 | 351 | 76 | |
BC104929.1 | - | 2 - 3 | 276 | 76 | |
BC104931.1 | - | 2 - 3 | 276 | 76 |