The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_147148.2 | NP_671489.1 | 7 - 8 | 880 | 130 |
GenBank mRNA | BI791533.1 | - | 1 - 2 | 130 | |
M99422.1 | - | 7 - 8 | 838 | 130 |