Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLK3
Entrez Gene ID
354
Gene Name
kallikrein related peptidase 3
Gene Aliases
APS, KLK2A1, PSA, hK3
Chromosome Location
Chr.19: 50854915 - 50860764 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BF679511.1 - 2 - 2 371 125

View Full Product Details