The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304548.1 | NP_001291477.1 | 59 - 60 | 8765 | 60 |
GenBank mRNA | BX111076.1 | - | 4 - 5 | 374 | 60 |
CD106515.1 | - | 4 - 5 | 514 | 60 |