Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP47
Entrez Gene ID
286464
Gene Name
cilia and flagella associated protein 47
Gene Aliases
CHDC2, CXorf22, CXorf30, CXorf59
Chromosome Location
Chr.X: 35919734 - 36385319 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304548.1 NP_001291477.1 60 - 61 8920 148
GenBank mRNA BX111076.1 - 5 - 6 529 148
CD106515.1 - 5 - 6 669 148

View Full Product Details