Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNGR1
Entrez Gene ID
9145
Gene Name
synaptogyrin 1
Gene Aliases
-
Chromosome Location
Chr.22: 39349949 - 39385588 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004711.4 NP_004702.2 2 - 3 397 77
NM_145731.3 NP_663783.1 2 - 3 397 77
NM_145738.2 NP_663791.1 2 - 3 363 77
GenBank mRNA AJ002303.1 - 2 - 3 363 77
AJ002304.1 - 2 - 3 335 77
AJ002305.1 - 2 - 3 335 77
AK289507.1 - 2 - 3 344 77
AK295863.1 - 2 - 3 355 77
BC000731.2 - 2 - 3 361 77
BI667990.1 - 2 - 3 421 77
BT007135.1 - 2 - 3 335 77
CR456590.1 - 2 - 3 345 77

View Full Product Details