Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNORD36B
Entrez Gene ID
26814
Gene Name
small nucleolar RNA, C/D box 36B
Gene Aliases
RNU36B, U36b
Chromosome Location
Chr.9: 133350094 - 133350164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_000017.1 1 - 1 39 71

View Full Product Details