The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_001191016.2 | NP_001177945.2 | 2 - 3 | 506 | 86 | |
NR_034061.2 | 2 - 3 | 506 | 86 | |||
NR_034063.2 | 2 - 3 | 506 | 86 | |||
NR_034064.2 | 2 - 3 | 506 | 86 | |||
NR_034065.2 | 2 - 3 | 506 | 86 | |||
NR_034067.2 | 2 - 3 | 506 | 86 | |||
GenBank mRNA | AF464191.1 | - | LIT2097 CASP12 pseudogene |
2 - 3 | 506 | 86 |
AF464192.1 | - | 2 - 3 | 506 | 86 | ||
AF464193.1 | - | 2 - 3 | 506 | 86 | ||
AF464194.1 | - | 2 - 3 | 506 | 86 | ||
AF486846.1 | - | 2 - 3 | 506 | 86 | ||
AY358222.1 | - | 2 - 3 | 281 | 86 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC102723895 | LOC102723895 | -283.0kb | Sense | Chr. - 104568489 - 104609302 on Build GRCh38 | |
Downstream | caspase 4, apoptosis-related cysteine peptidase pseudogene | LOC643733 | 4.1kb | Sense | Chr. - 104901549 - 104918175 on Build GRCh38 |