Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TTTY20
- Entrez Gene ID
-
252951
- Gene Name
- testis-specific transcript, Y-linked 20 (non-protein coding)
- Gene Aliases
-
NCRNA00145
- Chromosome Location
-
Chr.Y: 9329880 - 9334832 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001546.1 |
|
1 - 2 |
79 |
75 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF527832.3 |
- |
1 - 2 |
79 |
|
Chr. Y - 9329880 - 9395788 [-] on Build GRCh38 |
75 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105379270 |
LOC105379270 |
|
|
Sense |
Chr. - 9394048 - 9396987 on Build GRCh38 |
Downstream |
testis specific protein, Y-linked 3 |
TSPY3 |
CT78 |
2.6kb
|
Anti-sense |
Chr. - 9398421 - 9401224 on Build GRCh38 |
View Full Product Details