Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SUMO1P1
- Entrez Gene ID
-
391257
- Gene Name
- SUMO1 pseudogene 1
- Gene Aliases
-
PIC1L, UBL2, UBL6
- Chromosome Location
-
Chr.20: 53874498 - 53875709 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002189.3 |
|
1 - 1 |
1050 |
164 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105372672 |
LOC105372672 |
|
-239.9kb
|
Anti-sense |
Chr. - 53608354 - 53634590 on Build GRCh38 |
Downstream |
breast carcinoma amplified sequence 1 |
BCAS1 |
AIBC1, NABC1 |
67.8kb
|
Sense |
Chr. - 53943538 - 54070765 on Build GRCh38 |
View Full Product Details