Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GEMIN8P4
- Entrez Gene ID
-
492303
- Gene Name
- gem nuclear organelle associated protein 8 pseudogene 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 89993265 - 89994966 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_002830.1 |
|
1 - 1 |
248 |
152 |
GenBank mRNA |
AY726571.1 |
- |
1 - 1 |
239 |
152 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CX761685.1 |
- |
1 |
270 |
|
Chr. 1 - 89994307 - 89994953 [-] on Build GRCh38 |
152 |
GenBankmRNA |
BM019253.1 |
- |
1 |
189 |
|
Chr. 1 - 89994285 - 89994907 [-] on Build GRCh38 |
152 |
GenBankmRNA |
BI828015.1 |
- |
|
249 |
|
Chr. - - [-] on Build |
152 |
GenBankmRNA |
BM543897.1 |
- |
|
259 |
|
Chr. - - [-] on Build |
152 |
GenBankmRNA |
BU159632.1 |
- |
1 |
245 |
|
Chr. 1 - 89994020 - 89994963 [-] on Build GRCh38 |
152 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
leucine rich repeat containing 8 family member D |
LRRC8D |
LRRC5 |
-56.8kb
|
Anti-sense |
Chr. - 89821014 - 89936432 on Build GRCh38 |
Downstream |
zinc finger protein 326 |
ZNF326 |
ZAN75, ZIRD, Zfp326, dJ871E2.1 |
153.0
|
Anti-sense |
Chr. - 89995095 - 90035531 on Build GRCh38 |
View Full Product Details