Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNORA84
Entrez Gene ID
100124534
Gene Name
small nucleolar RNA, H/ACA box 84
Gene Aliases
-
Chromosome Location
Chr.9: 92292461 - 92292593 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_003704.2 1 - 1 62 64


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 3651 MIR3651 mir-3651 Sense Chr. - 92292458 - 92292547 on Build GRCh38
Downstream nucleolar protein 8 NOL8 C9orf34, NOP132, bA62C3.3, bA62C3.4 4.8kb Sense Chr. - 92297358 - 92325594 on Build GRCh38

View Full Product Details