The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003945.1 | 1 - 1 | 1946 | 118 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial ribosomal protein L17 | MRPL17 | L17mt, LIP2, MRP-L17, MRP-L26, RPL17L, RPML26 | -29.7kb | Sense | Chr. - 6680385 - 6683401 on Build GRCh38 |
Downstream | olfactory receptor family 2 subfamily AG member 2 | OR2AG2 | OR11-76, OR2AG2P | 45.5kb | Sense | Chr. - 6768007 - 6768957 on Build GRCh38 |