Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RPL13AP3
- Entrez Gene ID
-
645683
- Gene Name
- ribosomal protein L13a pseudogene 3
- Gene Aliases
-
RPL13A_11_1370
- Chromosome Location
-
Chr.14: 55766245 - 55767717 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_004844.1 |
|
1 - 1 |
1349 |
92 |
GenBank mRNA |
BC067891.1 |
- |
1 - 1 |
1349 |
92 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
kinectin 1 |
KTN1 |
CG1, KNT, MU-RMS-40.19 |
-81.7kb
|
Sense |
Chr. - 55580207 - 55684584 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 520 |
LINC00520 |
C14orf34 |
13.4kb
|
Anti-sense |
Chr. - 55781135 - 55796674 on Build GRCh38 |
View Full Product Details