The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003558.1 | 1 - 1 | 48 | 83 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | GRB2 associated regulator of MAPK1 subtype 1 | GAREM1 | C18orf11, FAM59A, GAREM, Gm944 | -41.2kb | Anti-sense | Chr. - 32263521 - 32470518 on Build GRCh38 |
Downstream | kelch like family member 14 | KLHL14 | 158.0kb | Anti-sense | Chr. - 32672673 - 32773011 on Build GRCh38 |